Merck
  • Effects of cucurbitacin e, a tetracyclic triterpene compound from Cucurbitaceae, on the pharmacokinetics and pharmacodynamics of warfarin in rats.
Effects of cucurbitacin e, a tetracyclic triterpene compound from Cucurbitaceae, on the pharmacokinetics and pharmacodynamics of warfarin in rats.
Basic & clinical pharmacology & toxicology (2014-09-25)
Tonggui Ding, Yuanjin Zhang, Ang Chen, Yu Tang, Mingyao Liu, Xin Wang
ZUSAMMENFASSUNG
This study firstly investigated the effects of cucurbitacin E (CuE), a tetracyclic triterpene compound from Cucurbitaceae, on the pharmacokinetics (PK) and pharmacodynamics (PD) of warfarin, a model CYP2C probe substrate, in the rat. In PK studies, the concentration of warfarin in blood samples was determined by HPLC-DAD, and the PK parameters were analysed using non-compartmental methods. In PD studies, the prothrombin time (PT) in blood plasma at each sample point was measured via thromboplastin reagents. CuE treatment (50, 100 and 200 μg/kg, i.p.) decreased warfarin clearance (28-32%), increased the area under the curve (AUC0-∞; 55-62%) and prolonged plasma half-life (t1/2; 58-72%). At the same time, the anticoagulation effect of warfarin (PTmax) was also significantly increased in the presence of CuE. These data demonstrated that CuE affected the PK and PD of warfarin, and these effects may be due to the inhibition of CYP2C activity by CuE. Hence, careful monitoring should be carried out during concomitant use of herbal products containing CuE with drugs that are metabolized by CYP2C enzymes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Ethylacetat, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Ethylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG