Merck
  • Human cytomegalovirus miR-US33-5p inhibits viral DNA synthesis and viral replication by down-regulating expression of the host Syntaxin3.
Human cytomegalovirus miR-US33-5p inhibits viral DNA synthesis and viral replication by down-regulating expression of the host Syntaxin3.
FEBS letters (2015-01-15)
Xin Guo, Ying Qi, Yujing Huang, Zhongyang Liu, Yanping Ma, Yaozhong Shao, Shujuan Jiang, Zhengrong Sun, Qiang Ruan
ZUSAMMENFASSUNG
During infection with human cytomegalovirus (HCMV), overexpression of hcmv-miR-US33 can inhibit the lytic viral replication and down-regulate US29 mRNA. However, it remains unknown whether inhibition of viral replication by miR-US33 is mediated by down-regulation of expression of US29 or another host gene. Here, we identified the host gene Syntaxin3 (STX3) to be a direct target of hcmv-miR-US33-5p using Hybrid-PCR and luciferase-reporter assays. It was further demonstrated that the levels of STX3 protein were down-regulated in hcmv-miR-US33-5p-overexpressing cells. Experiments with STX3-specific siRNA, or with an inhibitor of hcmv-miR-US33-5p confirmed that hcmv-miR-US33-5p-mediated inhibition of HCMV DNA synthesis and of viral replication are specifically mediated by down-regulation of STX3 expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, Deionized
Supelco
Wasser
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Wasser, BioPerformance Certified
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Wasser, ACS reagent
Supelco
Wasser, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Wasser, endotoxin, free
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O