Merck
  • Quantitative neuropeptidomics study of the effects of temperature change in the crab Cancer borealis.
Quantitative neuropeptidomics study of the effects of temperature change in the crab Cancer borealis.
Journal of proteome research (2014-09-13)
Ruibing Chen, Mingming Xiao, Amanda Buchberger, Lingjun Li
ZUSAMMENFASSUNG
Temperature changes influence the reaction rates of all biological processes, which can pose dramatic challenges to cold-blooded organisms, and the capability to adapt to temperature fluctuations is crucial for the survival of these animals. In order to understand the roles that neuropeptides play in the temperature stress response, we employed a mass spectrometry-based approach to investigate the neuropeptide changes associated with acute temperature elevation in three neural tissues from the Jonah crab Cancer borealis. At high temperature, members from two neuropeptide families, including RFamide and RYamide, were observed to be significantly reduced in one of the neuroendocrine structures, the pericardial organ, while several orcokinin peptides were detected to be decreased in another major neuroendocrine organ, the sinus gland. These results implicate that the observed neuropeptides may be involved with temperature perturbation response via hormonal regulation. Furthermore, a temperature stress marker peptide with the primary sequence of SFRRMGGKAQ (m/z 1137.7) was detected and de novo sequenced in the circulating fluid (hemolymph) from animals under thermal perturbation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
2,5-Dihydroxybenzoesäure, 98%