Merck
  • Sphere-forming capacity as an enrichment strategy for epithelial-like stem cells from equine skin.
Sphere-forming capacity as an enrichment strategy for epithelial-like stem cells from equine skin.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2014-10-04)
Bizunesh M Borena, Evelyne Meyer, Koen Chiers, Ann Martens, Kristel Demeyere, Sarah Y Broeckx, Luc Duchateau, Jan H Spaas
ZUSAMMENFASSUNG
Mammal skin plays a pivotal role in several life preserving processes and extensive damage may therefore be life threatening. Physiological skin regeneration is achieved through ongoing somatic stem cell differentiation within the epidermis and the hair follicle. However, in severe pathological cases, such as burn wounds, chronic wounds, and ulcers, the endogenous repair mechanisms might be insufficient. For this reason, exogenous purification and multiplication of epithelial-like stem/progenitor cells (EpSCs) might be useful in the treatment of these skin diseases. However, only few reports are available on the isolation, purification and characterization of EpSCs using suspension cultures. In the present study, skin was harvested from 6 mares and EpSCs were isolated and purified. In addition to their characterization based on phenotypic and functional properties, sphere formation was assessed upon isolation, i.e. at passage 0 (P0), and at early (P4) and late (P10) passages using different culture conditions. On average 0.53 ± 0.28% of these primary skin-derived cells showed the capacity to form spheres and hence possessed stem cell properties. Moreover, significantly more spheres were observed in EpSC medium versus differentiation medium, corroborating the EpSCs' privileged ability to survive in suspension. Furthermore, the number of cells per sphere significantly increased over time as well as with subsequent passaging. Upon immunophenotyping, the presumed EpSCs were found to co-express cytokeratin (CK) 14, Casein kinase 2 beta and Major Histocompatibility Complex (MHC) I and expressed no pan CK and wide CK. Only a few cells expressed MHC II. Their differentiation towards keratinocytes (at P4 and P10) was confirmed based on co-expression of CK 14, Casein kinase 2 beta, pan CK and wide CK. In one of six isolates, a non-EpSC cell type was noticed in adherent culture. Although morphological features and immunohistochemistry (IHC) confirmed a keratinocyte phenotype, this culture could be purified by seeding the cells in suspension at ultralow clonal densities (1 and 10 cells/cm(2)), yet with a significantly lower sphere forming efficiency in comparison to pure EpSCs (P = 0.0012). The present study demonstrated sphere formation as a valuable tool to purify EpSCs upon their isolation and assessed its effectiveness at different clonal seeding densities for eliminating a cellular contamination.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Supelco
Wasserstoffperoxid -Lösung, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard