Merck
  • Dominance of CTX-M-type extended-spectrum β-lactamase (ESBL)-producing Escherichia coli isolated from patients with community-onset and hospital-onset infection in China.
Dominance of CTX-M-type extended-spectrum β-lactamase (ESBL)-producing Escherichia coli isolated from patients with community-onset and hospital-onset infection in China.
PloS one (2014-07-02)
Shu Xia, Xin Fan, Zengguang Huang, Liang Xia, Meng Xiao, Rongchang Chen, Yingchun Xu, Chao Zhuo
ZUSAMMENFASSUNG
To investigate CTX-M genotypes among extended-spectrum β-lactamase-producing Escherichia coli (ESBL-EC) isolated from patients with community-onset and hospital-onset infections in China, their clonality and the distribution of CTX-M variants in different specimens of community-onset and hospital-onset infections. ESBL-EC isolates were collected from general hospitals from 2011 to 2012 in China. Broth microdilution method antimicrobial susceptibility testing of 16 antibiotics was performed. Clinical data from community-onset and hospital-onset infections due to ESBL-EC were analyzed. ESBL-encoding genes were amplified by PCR and sequenced, and multilocus sequence typing (MLST) was performed for a random selection of predominant CTX-M type strains identified. A total of 1,168 ESBL-EC isolates were obtained from various clinical specimens, 41.7% of which were responsible for causing community-onset infections. The presence of urinary calculi was higher in community-onset infections, whereas malignancy, cardiovascular and cerebrovascular diseases, dementia, chronic renal disease, diabetes mellitus and surgical treatment were found to have higher proportions in hospital-onset infections. There was no significant difference in trauma between community-onset and hospital-onset infections. 96.2% of the isolates were detected to harbor blaCTX-M genes. blaCTX-M-1 group and blaCTX-M-9 group were detected at 40.7% and 48.7% respectively, and both positive group accounted for 10.6%. blaCTX-M-55 (24.8%) and blaCTX-M-15 (18.2%) were the major genotypes in blaCTX-M-1 group while blaCTX-M-14 (46.8%) was predominant in blaCTX-M-9 group. A comparison of blaCTX-M distribution in different specimens between ESBL-EC causing community-onset and hospital-onset infection showed no significant difference. A total of 229 isolates were tested for MLST. ST131 (14%) was the predominant type. ST648, ST405 and ST1193 were also detected. Community-onset ESBL-EC has emerged as a common pathogen in China. CTX-M-14 is the most commonly encountered, CTX-M-55 and CTX-M-15 have spread rapidly. ST131 is the predominant clonal group, and the great diversity of CTX-M-producing isolates of E. coli has emerged in China.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sulbactam
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sulbactam, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sulbactam für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.