Merck
Plasma adipokine levels in Thais.
Asian Pacific journal of allergy and immunology (2015-04-05)
Preeya Leelahagul, Somchai Bovornkitti
ZUSAMMENFASSUNG
To assess three plasma adipokines that might act as biomarkers useful in determining persons exposed to asbestos. Sixty subjects were included in the study, 30 male workers from factories manufacturing cement products using chrysotile asbestos, and 30 volunteer controls comprising 15 men and 15 women. Fasting venous blood specimens were submitted for blood chemistry examination and analysis of plasma levels of three adipokines, i.e. adipsin, adiponectin and resistin. (1) Compared with non-obese subjects, the adiponectin levels were lower in obese subjects, resistin levels were higher, and hemoglobin concentrations were lower. (2) Adipsin levels in the workers were significantly higher than in the controls (p < 0.005), and this difference was not related to body fat. (3) There were no statistically significant differences in adiponectin, resistin, and adipsin levels between the male and female controls. (4) Adiponectin in the male controls was significantly higher than that in the male workers (p <0.05). (5) Female controls had significantly higher percentages of body fat (p < 0.0005) and resistin (p < 0.02) levels than male controls and male workers. (6) A significant negative correlation existed between resistin and hemoglobin levels levels (r= -0.336, p < 0.01). (7) Overall adipsin levels among male workers were significantly higher than among control subjects (p < 0.005); the six workers in whom asbestos bodies (AB) were detected did not have significantly higher levels of adipsin than those of workers without AB having been detected. Although the findings apparently showed higher adipsin levels in the workers, its value as a biomarker for asbestos exposure requires confirmation from studies on a larger group of subjects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Supelco
Saccharose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Saccharose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Harnsäure, ≥99%, crystalline
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Saccharose, ACS reagent
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Harnsäure, BioXtra, ≥99% (HPLC)
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)