Merck
Protein Drug Targets of Lavandula angustifolia on treatment of Rat Alzheimer's Disease.
Iranian journal of pharmaceutical research : IJPR (2015-01-07)
Hakimeh Zali, Mona Zamanian-Azodi, Mostafa Rezaei Tavirani, Alireza Akbar-Zadeh Baghban
ZUSAMMENFASSUNG
Different treatment strategies of Alzheimer's disease (AD) are being studied for treating or slowing the progression of AD. Many pharmaceutically important regulation systems operate through proteins as drug targets. Here, we investigate the drug target proteins in beta-amyloid (Aβ) injected rat hippocampus treated with Lavandula angustifolia (LA) by proteomics techniques. The reported study showed that lavender extract (LE) improves the spatial performance in AD animal model by diminishing Aβ production in histopathology of hippocampus, so in this study neuroprotective proteins expressed in Aβ injected rats treated with LE were scrutinized. Rats were divided into three groups including normal, Aβ injected, and Aβ injected that was treated with LE. Protein expression profiles of hippocampus tissue were determined by two-dimensional electrophoresis (2DE) method and dysregulated proteins such as Snca, NF-L, Hspa5, Prdx2, Apoa1, and Atp5a1were identified by MALDI-TOF/TOF. KEGG pathway and gene ontology (GO) categories were used by searching DAVID Bioinformatics Resources. All detected protein spots were used to determine predictedinteractions with other proteins in STRING online database. Different isoforms of important protein, Snca that exhibited neuroprotective effects by anti-apoptotic properties were expressed. NF-L involved in the maintenance of neuronal caliber. Hspa5 likewise Prdx2 displays as anti-apoptotic protein that Prdx2 also involved in the neurotrophic effects. Apoa1 has anti-inflammatory activity and Atp5a1, produces ATP from ADP. To sum up, these proteins as potential drug targets were expressed in hippocampus in response to effective components in LA may have therapeutic properties for the treatment of AD and other neurodegenerative diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Jodacetamid, BioUltra
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
MES -Lösung, BioUltra, Molecular Biology, 0.5 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Harnstoff, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Jodacetamid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Harnstoff, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat Hydrat, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Millipore
Harnstoff -Lösung, suitable for microbiology, 40% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Harnstoff, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
ATPA, solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.