Merck
Antitumor effects of baculovirus-infected dendritic cells against human pancreatic carcinoma.
Gene therapy (2014-07-11)
A Fujihira, T Suzuki, M O Chang, T Moriyama, M Kitajima, H Takaku
ZUSAMMENFASSUNG
Recently, we showed that baculovirus (BV)-infected dendritic cells (DCs) (BV-DCs) induced antitumor immunity against established tumors in mice. These antitumor effects were CD8(+) T-cell and natural killer (NK) cell dependent but CD4(+) T-cell independent. In the current study, we examined the antitumor effect of BV-DCs on human pancreatic cancer cells (AsPC-1). After treatment with BV-infected bone marrow-derived dendritic cells (BMDCs), human pancreatic tumors caused by AsPC-1 cells in a nude mouse model were significantly reduced in size, and the survival of the mice was improved compared with that of non-immature BMDC (iDC)- and BV-DC-immunized mice. We also found that wild-type BV could activate human DCs (HDCs) and that NK cells were activated by BV-infected HDCs (BHDCs). Our findings show that BV-DCs can induce antitumor immunity, which paves the way for the use of this technique as an effective tool for DC immunotherapy against malignancies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland