Merck
  • Noninvasive monitoring of plasma L-dopa concentrations using sweat samples in Parkinson's disease.
Noninvasive monitoring of plasma L-dopa concentrations using sweat samples in Parkinson's disease.
Clinica chimica acta; international journal of clinical chemistry (2015-01-17)
Makoto Tsunoda, Masaaki Hirayama, Takao Tsuda, Kinji Ohno
ZUSAMMENFASSUNG
L-dopa (l-3,4-dihydroxyphenylalanine) is commonly used for treating Parkinson's disease (PD). However, regardless of its prominent effect, therapeutic range of L-dopa narrows down with disease progression, which leads to development of motor complications including wearing off and dyskinesias. In addition, intestinal absorption of L-dopa is inversely correlated with the amount of oral protein intake, and shows intra- and inter-day variability. Hence, frequent monitoring of plasma L-dopa concentrations is beneficial, but frequent venipuncture imposes physical and psychological burdens on patients with PD. We investigated the usefulness of sweat samples instead of plasma samples for monitoring L-dopa concentrations. With a monolithic silica disk-packed spin column and the high-performance liquid chromatography-electrochemical detection system, L-dopa in sweat samples was successfully quantified and analyzed in 23 PD patients. We found that the Pearson's correlation coefficient of the plasma and sweat l-dopa concentrations was 0.678. Although the disease durations and severities were not correlated with the deviation of the actual sweat L-dopa concentrations from the fitted line, acquisition of the sweat samples under a stable condition was technically difficult in severely affected patients. The deviations may also be partly accounted for by skin permeability of L-dopa. Measuring L-dopa concentrations in sweat is suitable to get further insights into the L-dopa metabolism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Levodopa, European Pharmacopoeia (EP) Reference Standard