Merck
  • Fenofibrate causes elevation of betaine excretion but not excretion of other osmolytes by healthy adults.
Fenofibrate causes elevation of betaine excretion but not excretion of other osmolytes by healthy adults.
Journal of clinical lipidology (2014-08-12)
Michael Lever, Christopher J McEntyre, Peter M George, Sandy Slow, Stephen T Chambers, Christelle Foucher
ZUSAMMENFASSUNG
Cross-sectional data suggest that bezafibrate increases betaine excretion in dyslipidemic patients. We aimed to demonstrate that fenofibrate induces increased betaine excretion in normal subjects and explore whether other 1-carbon metabolites and osmolytes are similarly affected. Urine was collected from 26 healthy adults before and after treatment with fenofibrate (145 mg/day for 6 weeks). Excretions of betaine, N,N-dimethylglycine, free choline, myo-inositol, taurine, trimethylamine-N-oxide, carnitine, and acetylcarnitine were measured by liquid chromatography with mass spectrometric detection. Fenofibrate increased the median betaine excretion from 7.5 to 25.8 mmol/mole creatinine (median increase 3-fold), P < .001. The median increase in N,N-dimethylglycine excretion was 2-fold (P < .001). Median choline excretion increased 12% (significant, P = .029). Participants with higher initial excretions tended to have larger increases (P < .001 in all 3 cases). Fenofibrate did not significantly change the median excretions of myo-inositol, taurine, trimethylamine-N-oxide, and carnitine. The excretion of acetylcarnitine decreased 4-fold on treatment, with no correlation between the baseline and after-treatment excretions. Changes in all urine components tested, except trimethylamine-N-oxide, positively correlated with changes in betaine excretion even when the median excretions before and after were not significantly different. Fibrates increase betaine, and to a lesser extent N,N-dimethylglycine and choline, excretion. Other osmolytes are not elevated. Because the increase in betaine excretion depends on the baseline excretion, large increases in excretion in the metabolic syndrome and diabetes (where baseline excretions are high) could be expected. Replacement with betaine supplements may be considered.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Taurin, ≥99%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
myo-Inositol, ≥99% (GC), BioReagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Taurin, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Trimethylamin-N-oxid, 95%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Cadmiumchlorid, 99.99% trace metals basis