Merck
  • Antioxidant potential, DNA protection, and HPLC-DAD analysis of neglected medicinal Jurinea dolomiaea roots.
Antioxidant potential, DNA protection, and HPLC-DAD analysis of neglected medicinal Jurinea dolomiaea roots.
BioMed research international (2014-07-02)
Naseer Ali Shah, Muhammad Rashid Khan, Kiran Naz, Mubarak Ali Khan
ZUSAMMENFASSUNG
Jurinea dolomiaea Boiss., family Compositae, is a medicinally important plant of alpine region. Its tuberous roots are used in various ailments in folk medicine. This study was undertaken to estimate total phenolic (TPC) and total flavonoid contents (TFC) and to determine anti-free radical potential by diverse in vitro antioxidant assays. Crude methanol extract (JDME) was fractionated into n-hexane (JDHE), chloroform (JDCE), ethyl acetate (JDEE), n-butanol (JDBE), and aqueous (JDAE) fractions. The results indicated that JDEE and JDCE constituted the highest amount of TFC (807 ± 7.2 mg rutin equivalent/g sample) and TPC (757 ± 9.4 mg gallic acid equivalent/g sample), respectively. Significant correlation of TFC with IC50 values was recorded for (•)OH (R (2) = 0.91), H2O2 (R (2) = 0.82), and ABTS (R (2) = 0.82) assay. It could be made clear that JDEE was the most potent in antioxidant activity as compared to others, with generally lower IC50 values for DPPH (41.1 ± 1.0  μ g/mL), ABTS (46.7 ± 0.6  μ g/mL), H2O2 (42.2 ± 0.9  μ g/mL), (•)OH (61.1 ± 1.1  μ g/mL), O2 (-) (152 ± 1.1  μ g/mL), and antilipid peroxidation (54.3 ± 1.6  μ g/mL). HPLC chromatogram of JDEE revealed the presence of catechin, caffeic acid, and rutin. The results indicated the antioxidant activities of J. dolomiaea roots and merit further investigations for their use in oxidative stress related disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Supelco
Essigsäure, analytical standard