Merck
  • Prediction of the zeta potentials and ionic descriptors of a silica hydride stationary phase with mobile phases of different pH and ionic strength.
Prediction of the zeta potentials and ionic descriptors of a silica hydride stationary phase with mobile phases of different pH and ionic strength.
Analytica chimica acta (2015-01-28)
Chadin Kulsing, Yuanzhong Yang, Maria T Matyska, Joseph J Pesek, Reinhard I Boysen, Milton T W Hearn
ZUSAMMENFASSUNG
In this study, the zeta potentials of a silica hydride stationary phase (Diamond Hydride™) in the presence of different water-acetonitrile mixtures (from 0-80% (v/v) acetonitrile) of different ionic strengths (from 0-40mM) and pH values (from pH 3.0-7.0) have been investigated. Debye-Hückel theory was applied to explain the effect of changes in the pH and ionic strength of these aqueous media on the negative zeta potential of this stationary phase. The experimental zeta potentials of the Diamond Hydride™ particles as a function of acetonitrile content up to 50% (v/v) correlated (R(2)=0.998) with the predicted zeta potential values based on this established theory, when the values of the dissociation constant of all related species, as well as viscosity, dielectric constant and refractive index of the aqueous medium were taken into consideration. Further, the retention behavior of basic, acidic and neutral analytes was investigated under mobile phase conditions of higher pH and lower ionic strength. Under these conditions, the Diamond Hydride™ stationary phase surface became more negative, as assessed from the increasingly more negative zeta potentials, resulting in the ion exchange characteristics becoming more dominant and the basic analytes showing increasing retention. Ionic descriptors were derived from these chromatographic experiments based on the assumption that linear solvation energy relationships prevail. The results were compared with predicted ionic descriptors based on the different calculated zeta potential values resulting in an overall correlation of R(2)=0.888. These studies provide fundamental insights into the impact on the separation performance of changes in the zeta potential of the Diamond Hydride™ surface with the results relevant to other silica hydride and, potentially, to other types of stationary phase materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
Ammoniumformat, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar