Merck
  • Knockdown of angiopoietin like-2 protects against angiotensin II-induced cerebral endothelial dysfunction in mice.
Knockdown of angiopoietin like-2 protects against angiotensin II-induced cerebral endothelial dysfunction in mice.
American journal of physiology. Heart and circulatory physiology (2014-12-21)
Carol Yu, Xiaoyan Luo, Natacha Duquette, Nathalie Thorin-Trescases, Eric Thorin
ZUSAMMENFASSUNG
Angiopoietin like-2 (angptl2) is a circulating pro-inflammatory and pro-oxidative protein, but its role in regulating cerebral endothelial function remains unknown. We hypothesized that in mice knockdown (KD) of angptl2, cerebral endothelial function would be protected against ANG II-induced damage. Subcutaneous infusion of ANG II (200 ng·kg(-1)·min(-1), n = 15) or saline (n = 15) was performed in 20-wk-old angptl2 KD mice and wild-type (WT) littermates for 14 days. In saline-treated KD and WT mice, the amplitude and the sensitivity of ACh-induced dilations of isolated cerebral arteries were similar. However, while endothelial nitric oxide (NO) synthase (eNOS)-derived O2 (-)/H2O2 contributed to dilation in WT mice, eNOS-derived NO (P < 0.05) was involved in KD mice. ANG II induced cerebral endothelial dysfunction only in WT mice (P < 0.05), which was reversed (P < 0.05) by either N-acetyl-l-cysteine, apocynin, gp91ds-tat, or indomethacin, suggesting the contribution of reactive oxygen species from Nox2 and Cox-derived contractile factors. In KD mice treated with ANG II, endothelial function was preserved, likely via Nox-derived H2O2, sensitive to apocynin and PEG-catalase (P < 0.05), but not to gp91ds-tat. In the aorta, relaxation similarly and essentially depended on NO; endothelial function was maintained after ANG II infusion in all groups, but apocynin significantly reduced aortic relaxation in KD mice (P < 0.05). Protein expression levels of Nox1/2 in cerebral arteries were similar among all groups, but that of Nox4 was greater (P < 0.05) in saline-treated KD mice. In conclusion, knockdown of angptl2 may be protective against ANG II-induced cerebral endothelial dysfunction; it favors the production of NO, likely increasing endothelial cell resistance to stress, and permits the expression of an alternative vasodilatory Nox pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Anti--α-Glattmuskel-Aktin-Antikörper, Maus, monoklonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Supelco
Acrylamid, analytical standard
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Acetovanillon, ≥98%, FG
Sigma-Aldrich
4′-Hydroxy-3′-Methoxyacetophenon, 98%
Sigma-Aldrich
Acrylamid, purum, ≥98.0% (GC)
Sigma-Aldrich
Nω-Nitro-L-Arginin, ≥98% (TLC)
Supelco
Acrylamid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis