Merck
  • EphA2 promotes epithelial-mesenchymal transition through the Wnt/β-catenin pathway in gastric cancer cells.
EphA2 promotes epithelial-mesenchymal transition through the Wnt/β-catenin pathway in gastric cancer cells.
Oncogene (2013-06-12)
J Huang, D Xiao, G Li, J Ma, P Chen, W Yuan, F Hou, J Ge, M Zhong, Y Tang, X Xia, Z Chen
ZUSAMMENFASSUNG
This study aims to investigate the significance of erythropoietin-producing hepatocellular (Eph)A2 expression and the mechanism by which EphA2 is involved in the epithelial-mensenchymal transition (EMT) of gastric cancer cells. EphA2 expression levels were upregulated and positively correlated with metastasis and EMT markers in human gastric cancer specimens. Modulation of EphA2 expression levels had distinct effects on cell proliferation, cell cycle, migration, invasion and morphology in the gastric cancer cell lines SGC7901 and AGS in vitro and in vivo. Overexpression of EphA2 resulted in the upregulation of the EMT molecular markers N-cadherin and Snail, as well as the Wnt/β-catenin targets TCF4, Cyclin-D1 and c-Myc, while silencing EphA2 using short hairpin RNA had the opposite effect. Furthermore, inhibition of the Wnt/β-catenin pathway by XAV939 negated the effect of EphA2 overexpression, whereas activation of the Wnt/β-catenin pathway by LiCl impaired the effect of the EphA2 knockdown on EMT. These observations demonstrate that EphA2 upregulation is a common event in gastric cancer specimens that is closely correlated with cancer metastasis and that EphA2 promotes EMT of gastric cancer cells through activation of Wnt/β-catenin signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Lithiumchlorid, ACS reagent, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Lithiumchlorid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Lithiumchlorid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Lithiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
Elektrolyt -Lösung, nonaqueous, 2 M LiCl in ethanol
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Supelco
Lithiumchlorid -Lösung, 1 M in ethanol
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Supelco
Elektrolytlösung, nonaqueous, LiCl in ethanol (saturated)
Sigma-Aldrich
Lithiumchlorid, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Lithiumchlorid, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)