Merck
  • Hypericin damages the ectatic capillaries in a Roman cockscomb model and inhibits the growth of human endothelial cells more potently than hematoporphyrin does through induction of apoptosis.
Hypericin damages the ectatic capillaries in a Roman cockscomb model and inhibits the growth of human endothelial cells more potently than hematoporphyrin does through induction of apoptosis.
Photochemistry and photobiology (2014-07-30)
Zhuo-heng Li, De-sheng Meng, Yuan-yuan Li, Lai-chun Lu, Cai-ping Yu, Qian Zhang, Hai-yan Guan, Chen-wen Li, Xue Yang, Ruo-qiu Fu
ZUSAMMENFASSUNG
Hypericin (HY) is a promising photosensitizer (PS) for use in photodynamic therapy (PDT). Port-wine stains (PWSs) are congenital superficial dermal capillary malformations. In this study, we evaluated the photocytotoxic effects of HY for PDT in human vascular endothelial cells and a chicken cockscomb model. HY significantly inhibited the growth of human umbilical vein endothelial cells (HUVECs), as determined by colorimetric assays and morphological observation, and flow cytometry assays indicated induction of apoptosis and collapse of the mitochondrial membrane potential. In addition, HY more effectively inhibited growth of and induced apoptosis in HUVECs compared with hematoporphyrin (HP). Further experiments performed in a Roman chicken cockscomb model also showed a clear photocytotoxic effect on the cockscomb dermal capillary upon intravenous injection of HY. This effect may be due to the role of HY in the induction of apoptosis. Transmission electron microscopical analysis showed mitochondrial morphological changes such as incomplete ridges and swelling, and immunohistochemical assays showed an increase in the release of cytochrome c. In conclusion, HY exhibited a greater photocytotoxic activity than did HP toward the growth of endothelial cells and may thus represent a potent PS for PWS PDT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®