Merck
  • Solidification of floating organic drop liquid-phase microextraction cell fishing with gas chromatography-mass spectrometry for screening bioactive components from Amomum villosum Lour.
Solidification of floating organic drop liquid-phase microextraction cell fishing with gas chromatography-mass spectrometry for screening bioactive components from Amomum villosum Lour.
Biomedical chromatography : BMC (2014-09-25)
Xue Xue, Depo Yang, Dongmei Wang, Xinjun Xu, Longping Zhu, Zhimin Zhao
ZUSAMMENFASSUNG
In this study, a novel solidification of floating organic drop liquid-phase microextraction cell fishing with gas chromatography-mass spectrometry (SFOD-LPME-CF-GC-MS) method was established and used to screen, isolate and analyze bioactive components from Amomum villosum Lour. extract. Through comparision of its effect on the models of normal cell and inflammatory cells, anti-inflammatory active components of essential oil from A. villosum Lour. were readily screened, and the components obtained are in agreement with related pharmacological articles. SFOD-LPME-CF-GC-MS was used to analyze the interaction of A. villosum Lour. extracts with normal and lipopolysaccharide-stimulated RAW264.7 macrophage cells. The effect of A. villosum Lour. essential oil extracts in the LPS-stimulated RAW264.7 model were also assessed in terms of cytotoxicity and nitric oxide production as an indication of bioactivity. Three potentially bioactive components were identified, demonstrating that SFOD-LPME-CF-GC-MS can be used successfully in the drug-screening process. This approach avoids the requirement for protein precipitation, but more importantly, generates a high concentration ratio, allowing analysis of trace components in traditional Chinese medicines. SFOD-LPME-CF-GC-MS is a simple, fast, effective and reliable method for the screening and analysis of bioactive components, and it can be extended to screen other bioactive components from TCMs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfid, ≥99%, FCC, FG
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Dimethylsulfid, ≥99%
Sigma-Aldrich
Dimethylsulfid, anhydrous, ≥99.0%
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
Dimethylsulfid, natural, ≥99%, FCC, FG
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Supelco
Dimethylsulfid, analytical standard
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®