Merck
  • Protease-mediated release of chemotherapeutics from mesoporous silica nanoparticles to ex vivo human and mouse lung tumors.
Protease-mediated release of chemotherapeutics from mesoporous silica nanoparticles to ex vivo human and mouse lung tumors.
ACS nano (2015-02-24)
Sabine H van Rijt, Deniz A Bölükbas, Christian Argyo, Stefan Datz, Michael Lindner, Oliver Eickelberg, Melanie Königshoff, Thomas Bein, Silke Meiners
ZUSAMMENFASSUNG
Nanoparticles allow for controlled and targeted drug delivery to diseased tissues and therefore bypass systemic side effects. Spatiotemporal control of drug release can be achieved by nanocarriers that respond to elevated levels of disease-specific enzymes. For example, matrix metalloproteinase 9 (MMP9) is overexpressed in tumors, is known to enhance the metastatic potency of malignant cells, and has been associated with poor prognosis of lung cancer. Here, we report the synthesis of mesoporous silica nanoparticles (MSNs) tightly capped by avidin molecules via MMP9 sequence-specific linkers to allow for site-selective drug delivery in high-expressing MMP9 tumor areas. We provide proof-of-concept evidence for successful MMP9-triggered drug release from MSNs in human tumor cells and in mouse and human lung tumors using the novel technology of ex vivo 3D lung tissue cultures. This technique allows for translational testing of drug delivery strategies in diseased mouse and human tissue. Using this method we show MMP9-mediated release of cisplatin, which induced apoptotic cell death only in lung tumor regions of Kras mutant mice, without causing toxicity in tumor-free areas or in healthy mice. The MMP9-responsive nanoparticles also allowed for effective combinatorial drug delivery of cisplatin and proteasome inhibitor bortezomib, which had a synergistic effect on the (therapeutic) efficiency. Importantly, we demonstrate the feasibility of MMP9-controlled drug release in human lung tumors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
Triethylamin, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture