Merck
  • Determination of trace acrylamide in starchy foodstuffs by HPLC using a novel mixed-mode functionalized calixarene sorbent for solid-phase extraction cleanup.
Determination of trace acrylamide in starchy foodstuffs by HPLC using a novel mixed-mode functionalized calixarene sorbent for solid-phase extraction cleanup.
Journal of agricultural and food chemistry (2014-06-17)
Wenfen Zhang, Zhifen Deng, Wenjie Zhao, Ling Guo, Wei Tang, Huifang Du, Lin Lin, Qiong Jiang, Ajuan Yu, Lijun He, Shusheng Zhang
ZUSAMMENFASSUNG
In this paper, a rapid and effective HPLC method, using tetraazacalix[2]arene[2]triazine-modified silica gel (NCSi) as solid-phase extraction (SPE) sorbent, was developed for the purification and determination of trace acrylamide in starchy foodstuffs. The main influence factors of SPE including amount of NCSi sorbent, sample flow rate, and volume and composition of washing solution were investigated and evaluated in the sample pretreatment step. The optimized purification effect was achieved at the sample flow rate of 3 mL/min with 100 mg of NCSi and 2 mL of washing solution (water, 100%). The HPLC separation was carried out on a C18 column (250×4.6 mm i.d., 5 μm) with a mobile phase of methanol/water (10:90, v/v). The linear range of the calibration curve was 4-4000 ng/mL with s correlation coefficient of >0.9999. The intraday and interday RSDs (n=5) of peak areas of acrylamide were 0.22 and 0.90% and the intraday and interday RSDs (n=5) of retention times were 0.50 and 1.63%, respectively. In addition, overall recoveries through the extraction and NCSi-SPE purification ranged from 73.13 to 98%. Compared with the commercial SPE sorbents, NCSi featured excellent selectivity to retain polar and nonpolar interferences in the sample matrices. The improved method was simple, rapid, accurate, and promising for the determination of trace acrylamide in starchy foods with a complex matrix.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, analytical standard
Supelco
Hexan, analytical standard