Merck
  • Bioactive molecules as authenticity markers of Italian Chinotto (Citrus×myrtifolia) fruits and beverages.
Bioactive molecules as authenticity markers of Italian Chinotto (Citrus×myrtifolia) fruits and beverages.
Journal of pharmaceutical and biomedical analysis (2014-12-09)
Michele Protti, Francesco Valle, Ferruccio Poli, Maria Augusta Raggi, Laura Mercolini
ZUSAMMENFASSUNG
Chinotto (Citrus×myrtifolia) is a uncommon fruit belonging to the Citrus genus, mainly cultivated in small areas of the Italian territory, where the main use concerns the eponymous drink, marketed with the name of "Chinotto". The lack of information about this fruit highlights the usefulness of nutraceutical compound characterization, as well as the need to identify genuineness markers in derived commercial products. An analytical strategy based on SPE-HPLC-F was developed to identify and quantify different bioactive compounds in Chinotto (Citrus×myrtifolia) fruits and commercial beverages. The method was fully validated and successfully applied to the analysis of nutraceutical compounds in Chinotto fruits of Italian origin and in some Chinotto-based beverages, granting reliable and consistent data. The obtained results provided preliminary key information about the bioactive profiling of Citrus×myrtifolia and proved the suitability of the selected compounds as authenticity markers of derived commercial soft drinks.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Phosphorsäure -Lösung, 85 wt. % in H2O, FCC, FG
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phosphorsäure, BioUltra, ≥85% (T)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis