Merck
  • Stage specific activity of synthetic antimalarial endoperoxides, N-89 and N-251, against Plasmodium falciparum.
Stage specific activity of synthetic antimalarial endoperoxides, N-89 and N-251, against Plasmodium falciparum.
Parasitology international (2014-12-03)
Masayuki Morita, Takahiko Koyama, Hitomi Sanai, Akira Sato, Akiko Hiramoto, Araki Masuyama, Masatomo Nojima, Yusuke Wataya, Hye-Sook Kim
ZUSAMMENFASSUNG
We have reported that two endoperoxides, N-89 and N-251, synthesized in 2001, possess potent antimalarial activities. Aiming at their eventual use for curing malaria in humans, we have been investigating various aspects of their antimalarial actions. Here we show that N-89 and N-251 inhibit the growth of Plasmodium falciparum within human erythrocytes in vitro at its lifecycle stage 'trophozoite' specifically. It is known that artemisinin compounds, which are currently used for curing malaria, have other stage-specificities. Therefore, it is likely that the antimalarial mechanism of N-89 and N-251 differs from those of artemisinin compounds. As malaria parasites resistant to artemisinin-based combination therapy are currently emerging in some tropical regions, N-89 and N-251 are candidates for overcoming these new problems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Sorbit, ≥98% (GC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
D-Sorbit, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbit, 99% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
D-Sorbit, ≥98% (GC), Molecular Biology
Supelco
Sorbit, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sorbit, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
D-Sorbit, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sorbit, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sorbit F -Lösung, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Sorbit, ≥98% (GC), BioXtra
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
D-Sorbit, FCC, FG
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
D-Sorbit, liquid, tested according to Ph. Eur.
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®