Merck
  • Influenza vaccine effectiveness estimates for Western Australia during a period of vaccine and virus strain stability, 2010 to 2012.
Influenza vaccine effectiveness estimates for Western Australia during a period of vaccine and virus strain stability, 2010 to 2012.
Vaccine (2014-09-17)
Avram Levy, Sheena G Sullivan, Simone S Tempone, Kerry L M Wong, Annette K Regan, Gary K Dowse, Paul V Effler, David W Smith
ZUSAMMENFASSUNG
During 2010-2012 the strain composition of the influenza vaccine in the Southern Hemisphere did not change, but the circulating virus type/subtype did. We pooled data for these years from the Western Australian sentinel medical practice surveillance system for influenza to estimate vaccine effectiveness (VE) by influenza virus type and subtype. A case test-negative design was used with VE estimated as (1-odds ratio)×100%. There were 2182 patients included in the analysis across the 3 years studied. The predominant subtype was A/H1pdm09 in 2010 and 2011, and A/H3 in 2012. The overall adjusted VE estimate against all influenza for 2010-2012 was 51% (95% CI: 36, 63). Estimates were highest against A/H1pdm09 at 74% (95% CI: 47, 87), followed by 56% (95% CI: 33, 71) for influenza B and lowest against A/H3 at 39% (95% CI: 13, 57). When analyses were restricted to compare influenza-positive patients with patients who tested positive for a non-influenza virus, overall adjusted VE was 59% (95% CI: 39, 72). These results suggest moderate protection against influenza by vaccination in Western Australia over the period 2010-2012, and are consistent with findings from other settings.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aceton, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, histological grade, ≥99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Aceton, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Aceton, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, natural, ≥97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.