Merck
Ametantrone-based compounds as potential regulators of Tau pre-mRNA alternative splicing.
Organic & biomolecular chemistry (2014-11-06)
Gerard Artigas, Paula López-Senín, Carlos González, Núria Escaja, Vicente Marchán
ZUSAMMENFASSUNG
Tau pre-mRNA contains a stem-loop structure involved in the regulation of the alternative splicing of tau protein. We describe here a new family of Tau RNA ligands selected by dynamic combinatorial chemistry based on the combination of ametantrone with small RNA-binding molecules. The most promising compound results from derivatization of one of the side chains of the anthraquinone ring with the small aminoglycoside neamine through a short spacer. This compound binds the RNA target with a high affinity in a preferred binding site, in which the heteroaromatic moiety intercalates in the bulged region of the stem-loop and its side chains and neamine interact with the major groove of the RNA. Importantly, binding of this compound to mutated RNA sequences involved in the onset of some tauopathies such as FTDP-17 restores their thermodynamic stability to a similar or even higher levels than that of the wild-type sequence, thereby revealing its potential as a modulator of Tau pre-mRNA splicing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Wasser, Deionized
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Supelco
Wasser, for TOC analysis
Sigma-Aldrich
Wasser, BioPerformance Certified
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Ammoniumcitrat tribasisch, ≥97% (titration)
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Wasser, ACS reagent
Supelco
Wasser, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Wasser, tested according to Ph. Eur.
Sigma-Aldrich
2,4-Dihydroxybenzoesäure, 97%
Supelco
Wasser, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
2′,4′,6′-Trihydroxyacetophenon Monohydrat, 98%
Sigma-Aldrich
Wasser, endotoxin, free
Supelco
2′,4′,6′-Trihydroxyacetophenon Monohydrat, suitable for matrix substance for MALDI-MS, ≥99.5%
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Dichte Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O