Merck
  • In vitro human fecal microbial metabolism of Forsythoside A and biological activities of its metabolites.
In vitro human fecal microbial metabolism of Forsythoside A and biological activities of its metabolites.
Fitoterapia (2014-10-05)
Shihua Xing, Ying Peng, Mengyue Wang, Daofeng Chen, Xiaobo Li
ZUSAMMENFASSUNG
The present study aimed to investigate the metabolism of Forsythoside A (FTA) by human fecal bacteria to clarify the relationship between its intestinal metabolism and its pharmacological activities. FTA was incubated with human fecal microflora in vitro to investigate its metabolic process, and highly sensitive and specific ultra-performance liquid chromatography/quadrupole time-of-flight mass spectrometry (UPLC-Q-TOF/MS) was performed using MetaboLynx software for metabolite analysis. Caffeic acid (CA) and hydroxytyrosol (HT) were obtained by hydrolysis of FTA, and CA was further hydrogenated to form 3,4-dihydroxybenzenepropionic acid (DCA). The anticomplementary, antimicrobial and antiendotoxin activities of FTA and its metabolites by human fecal microflora were evaluated in vitro with a hemolysis assay, the agar disc-diffusion method, the MIC value and the gel clot LAL assay, respectively. The metabolites showed higher biological activity than FTA, especially HT and DCA. Orally administered FTA may be metabolized to HT and DCA, and the pharmacological effects of FTA may be dependent on intestinal bacterial metabolism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material