Merck
  • Quantification of folate metabolites in serum using ultraperformance liquid chromatography tandem mass spectrometry.
Quantification of folate metabolites in serum using ultraperformance liquid chromatography tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-06-01)
Xiuwei Wang, Ting Zhang, Xin Zhao, Zhen Guan, Zhen Wang, Zhiqiang Zhu, Qiu Xie, Jianhua Wang, Bo Niu
ZUSAMMENFASSUNG
Folate deficiency is considered a risk factor for many diseases such as cancer, congenital heart disease and neural tube defects (NTDs). There is a pressing need for more methods of detecting folate and its main metabolites in the human body. Here, we developed a simple, fast and sensitive ultraperformance liquid chromatography tandem mass spectrometry (UPLC/MS/MS) method for the simultaneous quantifications of folate metabolites including folic acid, 5-methyltetrahydrofolate (5-MeTHF), 5-formyltetrahydrofolate (5-FoTHF), homocysteine (Hcy), S-adenosylmethionine (SAM) and S-adenosylhomocysteine (SAH). The method was validated by determining the linearity (r(2)>0.998), sensitivity (limit of detection ranged from 0.05 to 0.200ng/mL), intra- and inter-day precision (both CV<6%) and recovery (each analyte was >90%). The total analysis time was 7min. Serum samples of NTD-affected pregnancies and controls from a NTD high-risk area in China were analyzed by this method, the NTD serum samples showed lower concentrations of 5-MeTHF (P<0.05) and 5-FoTHF (P<0.05), and higher concentrations of Hcy (P<0.05) and SAH (P<0.05) compared with serum samples from controls, consistent with a previous study. These results showed that the method is sensitive and reliable for simultaneous determination of six metabolites, which might indicate potential risk factors for NTDs, aid early diagnosis and provide more insights into the pathogenesis of NTDs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Folsäure, ≥97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Folsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Folsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
S-(5′-Adenosyl)-L-Homocystein, crystalline
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
DL-Homocystein, ≥95% (titration)
USP
Folsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)