Merck
  • Delivery of antisense oligonucleotides using poly(alkylene oxide)-poly(propylacrylic acid) graft copolymers in conjunction with cationic liposomes.
Delivery of antisense oligonucleotides using poly(alkylene oxide)-poly(propylacrylic acid) graft copolymers in conjunction with cationic liposomes.
Journal of controlled release : official journal of the Controlled Release Society (2014-09-07)
Lavanya Y Peddada, Olga B Garbuzenko, David I Devore, Tamara Minko, Charles M Roth
ZUSAMMENFASSUNG
The clinical application of gene silencing is hindered by poor stability and low delivery efficiency of naked oligonucleotides. Here, we present the in vitro and in vivo behaviors of a rationally designed, ternary, self-assembled nanoparticle complex, consisting of an anionic copolymer, cationic DOTAP liposome, and antisense oligonucleotide (AON). The multifunctional copolymers are based on backbone poly(propylacrylic acid) (PPAA), a pH-sensitive hydrophobic polymer, with grafted poly(alkylene oxides) (PAOs) varying in extent of grafting and PAO chemistry. The nanoparticle complexes with PPAA-g-PAO copolymers enhance antisense gene silencing effects in A2780 human ovarian cancer cells. A greater amount of AON is delivered to ovarian tumor xenografts using the ternary copolymer-stabilized delivery system, compared to a binary DOTAP/AON complex, following intraperitoneal injection in mice. Further, intratumoral injection of the nanoparticle complexes containing 1 mol% grafted PAO reduced tumoral bcl-2 expression by up to 60%. The data for complexes across the set of PAO polymers support a strong role for the hydrophilic-lipophilic balance of the graft copolymer in achieving serum stability and cellular uptake. Based upon these results, we anticipate that this novel nanoparticle delivery system can be extended to the delivery of plasmid DNA, siRNA, or aptamers for preclinical and clinical development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
N,N′-Diisopropylcarbodiimid, 99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
1-Hydroxybenzotriazol Hydrat, ≥97.0% dry basis (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
1-Hydroxybenzotriazol Hydrat, wetted with not less than 14 wt. % water, 98% dry basis
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
1-Hydroxybenzotriazol Hydrat, wetted with not less than 20 wt. % water, 97% dry basis
Sigma-Aldrich
N,N′-Diisopropylcarbodiimid, purum, ≥98.0% (GC)
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Diisopropylcarbodiimide solution, 1 M in THF