Merck
  • Enhanced miR-210 expression promotes the pathogenesis of endometriosis through activation of signal transducer and activator of transcription 3.
Enhanced miR-210 expression promotes the pathogenesis of endometriosis through activation of signal transducer and activator of transcription 3.
Human reproduction (Oxford, England) (2014-12-18)
M Okamoto, K Nasu, W Abe, Y Aoyagi, Y Kawano, K Kai, M Moriyama, H Narahara
ZUSAMMENFASSUNG
What are the roles of the microRNA miR-210-an miRNA that is up-regulated in endometriotic cyst stromal cells (ECSCs)-in the pathogenesis of endometriosis? Up-regulated miR-210 expression in ECSCs is involved in their proliferation, resistance to apoptosis and angiogenesis through signal transducer and activator of transcription (STAT) 3. In the pathogenesis of endometriosis, a number of roles for microRNAs (miRNAs) are becoming apparent. ECSCs and normal endometrial stromal cells (NESCs) were isolated from ovarian endometriotic tissues (patients aged 24-40 years undergoing salpingo-oophorectomy or evisceration for the treatment of ovarian endometriotic cysts, n = 10) and the eutopic endometrial tissues without endometriosis (premenopausal patients aged 35-45 years undergoing hysterectomies for subserousal leiomyoma, n = 13), respectively. We used a global gene expression microarray technique to identify downstream targets of miR-210, and we assessed the functions of miR-210 in the pathogenesis of endometriosis by using the miR-210-transfected NESCs. Gene expression microarray analysis revealed that one of the key target molecules of miR-210 is STAT3. In the NESCs, in comparison to the control, miR-210 transfection resulted in the induction of cell proliferation (P < 0.0005), the production of vascular endothelial cell growth factor (VEGF) (P < 0.0005) and the inhibition of apoptosis (P < 0.05) through STAT3 activation [increased levels of mRNA (P < 0.0005), and protein (P < 0.005)]. In the ECSCs, inhibitors of STAT3 inhibited the cell proliferation and VEGF production (P < 0.05), and induced the apoptosis of these cells (P < 0.05). The roles of aberrant miR-210 expression were investigated only in the stromal component of ectopic and eutopic endometrium. Control endometrial tissues were obtained from premenopausal patients who had subserosal leiomyoma and NESC gene expression patterns may be altered in these women. Furthermore, the effects of STAT3 inhibitors were evaluated only in ECSCs and not in NESCs. The present findings indicate that miR-210 induces NESCs to differentiate into the endometriotic phenotype and we speculate that up-regulated miR-210 expression in ECSCs is involved in the creation of the endometriosis-specific cellular dysfunctions through epigenetic mechanisms. The data indicate that STAT3 inhibitors may be promising candidates for the treatment of endometriosis. This work was supported in part by Grants-in-Aid for Scientific Research from the Japan Society for the Promotion of Science (no. 13237327 to K.N., no. 25861500 to Y.K. and no. 23592407 to H.N.). There are no conflicts of interest to declare.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethylsulfoxid, analytical standard