Merck
  • Alpha-lipoic acid protects against methylmercury-induced neurotoxic effects via inhibition of oxidative stress in rat cerebral cortex.
Alpha-lipoic acid protects against methylmercury-induced neurotoxic effects via inhibition of oxidative stress in rat cerebral cortex.
Environmental toxicology and pharmacology (2014-12-20)
Tian-Yao Yang, Zhao-Fa Xu, Wei Liu, Bin Xu, Yu Deng, Yue-Hui Li, Shu Feng
ZUSAMMENFASSUNG
MeHg is one of the environmental pollutants that lead to oxidative stress and an indirect excitotoxicity caused by altered glutamate (Glu) concentration. However, little was known of the interaction. Therefore, we developed a rat model of MeHg poisoning to explore its neurotoxic effects, and whether LA could attenuate MeHg-induced neurotoxicity. Seventy-two rats were randomly divided into four groups: control group, MeHg-treated groups (4 and 12μmol/kg), and LA pre-treatment group. Administration of the 12μmol/kg MeHg for 4 weeks significantly increased ROS formation that might be critical to aggravate oxidative damages in cerebral cortex. Meanwhile, Glu metabolism as well as GLAST and GLT-1 appeared to be disrupted by MeHg exposure. Pre-treatment of the 35μmol/kg LA significantly prevented MeHg-induced oxidative stress and Glu dyshomoestasis. In conclusion, findings indicated that MeHg could induce oxidative stress and Glu uptake/metabolism disorders in cerebral cortex, LA might antagonize these neurotoxic effects induced by MeHg.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Verflüssigtes Phenol, ≥89.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Supelco
Phenol -Lösung, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol -Lösung, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol -Lösung, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard