Merck
  • Minimally invasive direct coronary artery bypass improves late survival compared with drug-eluting stents in isolated proximal left anterior descending artery disease: a 10-year follow-up, single-center, propensity score analysis.
Minimally invasive direct coronary artery bypass improves late survival compared with drug-eluting stents in isolated proximal left anterior descending artery disease: a 10-year follow-up, single-center, propensity score analysis.
The Journal of thoracic and cardiovascular surgery (2014-02-14)
Umberto Benedetto, Shahzad G Raja, Rafik F B Soliman, Alberto Albanese, Anand Jothidasan, Charles D Ilsley, Mohamed Amrani
ZUSAMMENFASSUNG
Minimally invasive direct coronary artery bypass (MIDCAB) has been proposed to reduce surgical morbidity and improve long-term outcomes compared with stenting in the treatment of isolated proximal left anterior descending artery. However, the survival benefit from MIDCAB still needs to be demonstrated, in particular, because percutaneous coronary intervention with drug-eluting stents (DES-PCI) continues to be considered the initial treatment strategy. We conducted a 10-year follow-up, single-center, propensity score-matched MIDCAB versus DES-PCI comparison. A total of 1033 patients (303 MIDCAB and 730 DES-PCI) with isolated proximal left anterior descending disease were included. Propensity score matching was used to compare 303 pairs of MIDCAB and DES-PCI patients. MIDCAB and DES-PCI presented with comparable 30-day mortality (2 of 303 [0.6%] vs 1 of 303 [0.3%]; P=1.0). At 10 years, DES-PCI was associated with a 2.19-fold increased risk of late death (95% confidence interval, 1.15-4.17), a 2.0-fold increased risk of repeat revascularization (95% confidence interval, 1.20-3.47), and a 2.14-fold increased risk of the composite of death and repeat revascularization (95% confidence interval, 1.41-3.24). These findings strongly support a survival benefit from MIDCAB at long-term follow-up compared with DES-PCI in the treatment of isolated left anterior descending disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rapamycin aus Streptomyces hygroscopicus, ≥95% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.