Merck
Evaluation of degradation kinetics and physicochemical stability of tenofovir.
Drug testing and analysis (2014-05-13)
Vivek Agrahari, Sandeep Putty, Christiane Mathes, James B Murowchick, Bi-Botti C Youan
ZUSAMMENFASSUNG
Tenofovir (TFV) has been proven to prevent the transmission of the Human Immunodeficiency Virus (HIV) through the vagina. But, there is little information available about its stability under various storage and stress conditions. Hence, this study aimed to investigate the degradation behavior and physicochemical stability of TFV using liquid chromatography coupled mass spectrometry (LC-MS) and solid state X-ray diffraction (XRD) analyses. The LC-MS analysis was performed on a QTrap mass spectrometer with an enhanced mass spectrum (EMS) scan in positive mode. A reversed phase C18 column was used as the stationary phase. TFV exhibited degradation under acidic and alkaline hydrolytic conditions. The degradation products with m/z 289.2 and 170 amu have been proposed as 6-Hydroxy adenine derivative of TFV, and (2-hydroxypropan-2-yloxy) methylphosphonic acid, respectively. A pseudo-first-order degradation kinetic allowed for estimating the shelf-life, half-life, and time required for 90% degradation of 3.84, 25.34, and 84.22 h in acidic conditions, and 58.26, 384.49, and 1277.75 h in alkaline conditions, respectively. No significant degradation was observed at pH 4.5 (normal cervicovaginal pH) and oxidative stress conditions of 3% and 30% v/v hydrogen peroxide solutions. The shelf life of TFV powder at room temperature was 23 months as calculated by using an Arrhenius plot. The XRD pattern showed that the drug was stable and maintained its original crystallinity under the accelerated and thermal stress conditions applied. Stability analyses revealed that the TFV was stable in various stress conditions; however, formulation strategies should be implemented to protect it in strong acidic and alkaline environments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Natronlauge, 5.0 M
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%