Merck
  • A sensitive and rapid HPLC-MS/MS method for the quantitative determination of trace amount of bromocriptine in small clinical prolactinoma tissue.
A sensitive and rapid HPLC-MS/MS method for the quantitative determination of trace amount of bromocriptine in small clinical prolactinoma tissue.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-03-31)
Qingce Zang, Yang Liu, Jiuming He, Xiaofei Yue, Ruiping Zhang, Renzhi Wang, Zeper Abliz
ZUSAMMENFASSUNG
Usually, insufficient intratumoral concentration of therapeutic drugs is one of the reasons for tumor treatment failure. However, little is known about intratumoral distribution of bromocriptine in non-responding prolactinomas because of extremely low drug concentration and small prolactinoma tissue samples. In this study, a sensitive, rapid and high-throughput quantitative bioanalytical method has been established by using high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS) for the determination of bromocriptine at trace level in human prolactinoma tissue. As little as 20 mg (wet weight) tissue sample was required and total analysis time was 6 min in this method. The assay quantifies over a linear range of 50 fg/mg to 5 pg/mg, and has a 25 fg/mg limit of detection at a signal/noise ratio of 3. This validated method was successfully used to quantitatively determine bromocriptine in clinical post-operative bromocriptine-sensitive and -resistant prolactinomas. The results revealed bromocriptine concentration in resistant prolactinomas (0.49-1.25 pg/mg) was significantly higher than that in sensitive prolactinomas (0.057-0.47 pg/mg). These results provided direct evidence to demonstrate the reseaon for failure of bromocriptine treatment in some patients with prolactinoma was "intrinsic" tumor (cell) resistence, rather than insufficient drug concentration in tumor tissue. Additionaly, this HPLC-MS/MS method has been shown to be suitable for bromocriptine analysis in small amount tissue sample and could be adapted for therapeutic drug monitoring of other clinical medicine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material