Merck
Genotoxicity of rice bran oil extracted by supercritical CO2 extraction.
Biological & pharmaceutical bulletin (2014-12-03)
Jae-Suk Choi, Eun Jin Cheon, Tae-Uk Kim, Woi-Sook Moon, Joo-Wan Kim, Mi-Ryung Kim
ZUSAMMENFASSUNG
Rice bran oil extracted by supercritical CO2 extraction (RB-SCE) reportedly exhibits pharmacological activities such as antioxidant and in vivo hair growth-inducing effects. Such activities raise the possibility of the development of novel hair growth-inducing agents using RB-SCE. The aim of this study was to investigate the potential genotoxic effects of RB-SCE in three short-term mutagenicity assays (bacterial reverse mutation assay, in vitro mammalian chromosomal aberration test, and in vivo micronucleus assay). RB-SCE showed no genotoxicity in the bacterial reverse mutation assay up to 5000 mg/plate and in the in vivo micronucleus test up to 600 mg/kg body weight. However, at 120 µg/mL with S9 mix and 200 µg/mL without S9 mix RB-SCE showed significantly different genotoxicity than the negative control in the in vitro chromosome aberration test. The induction of chromosomal aberrations under the present conditions may have no biological significance. We have herein demonstrated that RB-SCE can be regarded as a non-genotoxic material based on the available in vivo and in vitro results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Benzo[a]pyren, ≥96% (HPLC)
Sigma-Aldrich
Cyclophosphamid Monohydrat, bulk package
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
4-Nitrochinolin N-Oxid, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Anthranilsäure, reagent grade, ≥98%
Sigma-Aldrich
2-Aminoanthracen, 96%
Supelco
Benzo[a]pyren, analytical standard, for environmental analysis
Sigma-Aldrich
Cyclophosphamid Monohydrat, ISOPAC®
Supelco
Benzo[a]pyren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Sigma-Aldrich
2-Nitrofluoren, 98%
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
Anthranilsäure, puriss. p.a., ≥99.5% (T)
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard