- Conjugated polymer dots for ultra-stable full-color fluorescence patterning.
Conjugated polymer dots for ultra-stable full-color fluorescence patterning.
Small (Weinheim an der Bergstrasse, Germany) (2014-07-23)
Kaiwen Chang, Zhihe Liu, Haobin Chen, Lan Sheng, Sean Xiao-An Zhang, Daniel T Chiu, Shengyan Yin, Changfeng Wu, Weiping Qin
PMID25048985
ZUSAMMENFASSUNG
Stable full-color fluorescence patterning are achieved by multicolor polymer-dot inks. The fluorescent patterns show extraordinary stability upon various treatments, offering a superior combination of bright fluorescence, excellent photostability, chemical resistance, and eco-friendship.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max™, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)