Merck
  • ACAT1-associated Late Endosomes/Lysosomes Significantly Improve Impaired Intracellular Cholesterol Metabolism and the Survival of Niemann-Pick Type C Mice.
ACAT1-associated Late Endosomes/Lysosomes Significantly Improve Impaired Intracellular Cholesterol Metabolism and the Survival of Niemann-Pick Type C Mice.
Acta histochemica et cytochemica (2014-09-16)
Masashi Kamikawa, XiaoFeng Lei, Yukio Fujiwara, Kazuchika Nishitsuji, Hiroshi Mizuta, Motohiro Takeya, Naomi Sakashita
ZUSAMMENFASSUNG
We previously demonstrated that macrophages exhibit endoplasmic reticulum fragmentation under cholesterol-rich conditions, which results in the generation of acyl-coenzyme A: cholesterol acyltransferase 1 (ACAT1)-associated late endosomes/lysosomes (ACAT1-LE). ACAT1-LE efficiently esterify free cholesterol in loco, even with abnormal egress of free cholesterol from late endosomes. Because impaired free cholesterol transport from late endosomes results in Niemann-Pick type C disease (NPC), the induction of ACAT1-LE is a potential therapeutic intervention for NPC. To examine the effects of ACAT1-LE induction on intracellular cholesterol metabolism, we incubated bone marrow-derived macrophages possessing NPC phenotype (npc1 (-/-)) with methyl-β-cyclodextrin-cholesterol complex (mβCD-cho), a cholesterol donor. Immunofluorescence confocal microscopy revealed that mβCD-cho treatment of npc1 (-/-) macrophages resulted in significant colocalization of signals from ACAT1 and lysosome-associated membrane protein 2, a late endosome/lysosome marker. npc1 (-/-) macrophages contained significant amounts of free cholesterol with negligible amounts of cholesteryl ester, while wild-type macrophages possessed the same amounts of both cholesterols. mβCD-cho treatment also induced marked restoration of cholesterol esterification activity. mβCD-cho administration in neonate npc1 (-/-) mice improved survival. These results indicate that ACAT1-LE induction in npc1 (-/-) mice corrects impaired intracellular cholesterol metabolism and that restoring cholesterol esterification improves prognosis of npc1 (-/-). These data suggest that ACAT1-LE induction is a potential alternative therapeutic strategy for NPC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture