Merck
Performance of different three-dimensional scaffolds for in vivo endochondral bone generation.
European cells & materials (2014-06-11)
W Yang, S K Both, G Jvm van Osch, Y Wang, J A Jansen, F Yang
ZUSAMMENFASSUNG
In the context of skeletal tissue development and repair, endochondral ossification has inspired a new approach to regenerate bone tissue in vivo using a cartilage intermediate as an osteoinductive template. The aim of this study was to investigate the behavior of mesenchymal stem cells (MSCs) in regard to in vitro cartilage formation and in vivo bone regeneration when combined with different three-dimensional (3D) scaffold materials, i.e., hydroxyapatite/tricalcium phosphate (HA/TCP) composite block, polyurethane (PU) foam, poly(lactic-co-glycolic acid)/poly(ε-caprolactone) electrospun fibers (PLGA/PCL) and collagen I gel. To this end, rat MSCs were seeded on these scaffolds and chondrogenically differentiated in vitro for 4 weeks followed by in vivo subcutaneous implantation for 8 weeks. Nonetheless, the quality and maturity of in vivo ectopic bone formation appeared to be scaffold/material-dependent. Eight weeks of implantation was not sufficient to ossify the entire PLGA/PCL constructs, albeit a comprehensive remodeling of the cartilage had occurred. For HA/TCP, PU and collagen I scaffolds, more mature bone formation with rich vascularity and marrow stroma development could be observed. These data suggest that chondrogenic priming of MSCs in the presence of different scaffold materials allows the establishment of reliable templates for generating functional endochondral bone tissue in vivo without using osteoinductive growth factors. The morphology and maturity of bone formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Natriumnitrit, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Natriumnitrit, ACS reagent, ≥97.0%
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Prolin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
USP
Ascorbinsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methylenblau, certified by the BSC
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis