Merck
  • Gene expression markers in the zebrafish embryo reflect a hepatotoxic response in animal models and humans.
Gene expression markers in the zebrafish embryo reflect a hepatotoxic response in animal models and humans.
Toxicology letters (2014-07-30)
Marja Driessen, Anne S Kienhuis, Alexa P Vitins, Jeroen L A Pennings, Tessa E Pronk, Evert-Jan van den Brandhof, Marianne Roodbergen, Bob van de Water, Leo T M van der Ven
ZUSAMMENFASSUNG
The zebrafish embryo (ZFE) is a promising non-rodent model in toxicology, and initial studies suggested its applicability in detecting hepatotoxic responses. Here, we hypothesize that the detailed analysis of underlying mechanisms of hepatotoxicity in ZFE contributes to the improved identification of hepatotoxic properties of new compounds and to the reduction of rodents used for screening. ZFEs were exposed to nine reference hepatotoxicants, targeted at induction of cholestasis, steatosis and necrosis, and two non-hepatotoxic controls. Histopathology revealed various specific morphological changes in the ZFE hepatocytes indicative of cell injury. Gene expression profiles of the individual compounds were generated using microarrays. Regulation of single genes and of pathways could be linked to hepatotoxic responses in general, but phenotype-specific responses could not be distinguished. Hepatotoxicity-associated pathways included xenobiotic metabolism and oxidoreduction related pathways. Overall analysis of gene expression identified a limited set of potential biomarkers specific for a common hepatotoxicity response. This set included several cytochrome P450 genes (cyp2k19, cyp4v7, cyp2aa3), genes related to liver development (pklr) and genes important in oxidoreduction processes (zgc:163022, zgc:158614, zgc:101858 and sqrdl). In conclusion, the ZFE model allows for identification of hepatotoxicants, without discrimination into specific phenotypes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®