Merck
  • Endocrine disrupting effects of dichlorodiphenyltrichloroethane analogues on gonadotropin hormones in pituitary gonadotrope cells.
Endocrine disrupting effects of dichlorodiphenyltrichloroethane analogues on gonadotropin hormones in pituitary gonadotrope cells.
Environmental toxicology and pharmacology (2014-05-13)
Jinghua Zhou, Ye Yang, Kang Xiong, Jing Liu
ZUSAMMENFASSUNG
It has been shown that exposure to dichlorodiphenyltrichloroethane (DDT) analogues leads to disharmony of follicle-stimulating hormone (FSH) and luteinizing hormone (LH). However, the effects and mechanisms of DDT analogues on the expression of gonadotropin genes (FSHβ, LHβ and Cgα), which is the rate-limiting step of FSH and LH biosynthesis, remain unknown. In this study, we assessed the effects of p,p'-DDT, o,p'-DDT, p,p'-dichlorodiphenyldichloroethylene (p,p'-DDE) and methoxychlor (MXC) on gonadotropin genes expression and hormones synthesis in gonadotrope cells. p,p'-DDT and MXC at test concentrations ranging from 10(-9) to 10(-7)mol/L, stimulated gonadotropin genes expression and hormones synthesis in a dose-dependent manner. The activation of extracellular signal-regulated kinase (ERK) was required for the induction of gonadotropin genes expression and hormones synthesis by p,p'-DDT or MXC exposure. This study showed for the first time that p,p'-DDT and MXC regulated gonadotropin genes expression and hormones synthesis through ERK pathway in gonadotrope cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
PD184352, ≥98% (HPLC)
Dimethylsulfoxid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®