Merck
On formulating ophthalmic emulsions.
Colloids and surfaces. B, Biointerfaces (2014-07-14)
Roozbeh Mafi, Cameron Gray, Robert Pelton, Howard Ketelson, James Davis
ZUSAMMENFASSUNG
The formulation of dilute, transparent ophthalmic emulsions (eye drops) with long shelf lives is a challenge because of the tendency of the emulsion droplets to aggregate, particularly in the presence of the water-soluble polymers typically used in eye drops. While many functions of eye drops, such as lubricity and residence time in the eye, are promoted by high concentrations of high molecular weight water-soluble polymers, emulsified lipids and drugs aggregate in the eye drop bottle if the polymer concentration is above the critical flocculation concentration (CFC). The purpose is to develop a simple approach to predict the CFC for polymers based on information readily available in the literature. High molecular weight guar was hydrolyzed to give a series of guar samples spanning a wide range of average molecular weights. The CFC values and critical viscosity concentrations were measured as functions guar properties, using electrophoresis, dynamic light scattering and rheology measurements. The higher the guar molecular weight, the lower was the CFC, the maximum concentration that can be tolerated in the eye drop formulation. The guar CFC values were approximately equal to the overlap concentrations where guar molecules start to overlap in solution. We propose that the CFC can be estimated for any water-soluble polymer using the polymer molecular weight and the readily available Mark-Houwink parameters, thus providing a design rule for ophthalmic emulsions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Hexadecan, ReagentPlus®, 99%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Hexadecan, anhydrous, ≥99%
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Supelco
Hexadecan, analytical standard
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%