Merck
  • Enhanced oral bioavailability of morin administered in mixed micelle formulation with PluronicF127 and Tween80 in rats.
Enhanced oral bioavailability of morin administered in mixed micelle formulation with PluronicF127 and Tween80 in rats.
Biological & pharmaceutical bulletin (2015-03-10)
Yeon Ah Choi, You Hyun Yoon, Kwangik Choi, Mihwa Kwon, Soo Hyeon Goo, Jin-Sun Cha, Min-Koo Choi, Hye Suk Lee, Im-Sook Song
ZUSAMMENFASSUNG
To overcome the low oral bioavailability of morin, a mixed micelle formulation with pharmaceutical excipients that facilitate solubilization and modulate P-glycoprotein (P-gp) was developed and evaluated in vitro and in vivo rats. Morin-loaded mixed micelle formulation with a morin-PluronicF127-Tween80 ratio of 1 : 10 : 0.02 (w/w/w) was prepared by a thin-film hydration method. The solubility, size distribution, drug encapsulation efficiency, and percent drug loading of the formulation were characterized. Subsequently, in vivo pharmacokinetic parameters of morin loaded in a PluronicF127 and Tween80 mixed-micelle formulation were investigated in rats. Absolute bioavailability of morin was dramatically increased by the oral administration of morin-loaded PluronicF127 and Tween80 mixed micelle from 0.4% to 11.2% without changing the systemic clearance and half-life. In Caco-2 cells, absorption permeability of morin from the novel formulation was increased 3.6-fold compared with that of morin alone. P-gp inhibition by cyclosporine A (CsA) increased absorptive permeability of morin 2.4-fold but decreased the efflux of morin by 52%, which was consistent with increased plasma concentration of morin in the pretreatment of CsA in rats. The morin formulation inhibited P-gp transport activity by 83.1% at 100 µM as morin concentration. Moreover, morin formulation increased paracellular permeability of Lucifer yellow by 1.6-1.8 fold. In conclusion, enhanced oral bioavailability of morin from morin-loaded PluronicF127 and Tween80 mixed micelle formulation can be attributed to increased intestinal permeation of morin, which was mediated at least by P-gp inhibition and enhanced paracellular route.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
SAFC
L-Glutamin
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Piperazin, ReagentPlus®, 99%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Piperazin, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Ethylendiamintetraessigsäure, ≥98.0% (KT)
Quercetin Dihydrat, primary reference standard
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O