Merck
  • Hematogenously disseminated Orientia tsutsugamushi-infected murine model of scrub typhus [corrected].
Hematogenously disseminated Orientia tsutsugamushi-infected murine model of scrub typhus [corrected].
PLoS neglected tropical diseases (2014-07-11)
Thomas R Shelite, Tais B Saito, Nicole L Mendell, Bin Gong, Guang Xu, Lynn Soong, Gustavo Valbuena, Donald H Bouyer, David H Walker
ZUSAMMENFASSUNG
Orientia tsutsugamushi, the etiologic agent of scrub typhus, is a mite-borne rickettsia transmitted by the parasitic larval stage of trombiculid mites. Approximately one-third of the world's population is at risk of infection with Orientia tsutsugamushi, emphasizing its importance in global health. In order to study scrub typhus, Orientia tsutsugamushi Karp strain has been used extensively in mouse studies with various inoculation strategies and little success in inducing disease progression similar to that of human scrub typhus. The objective of this project was to develop a disease model with pathology and target cells similar to those of severe human scrub typhus. This study reports an intravenous infection model of scrub typhus in C57BL/6 mice. This mouse strain was susceptible to intravenous challenge, and lethal infection occurred after intravenous inoculation of 1.25 × 10(6) focus (FFU) forming units. Signs of illness in lethally infected mice appeared on day 6 with death occurring ∼ 6 days later. Immunohistochemical staining for Orientia antigens demonstrated extensive endothelial infection, most notably in the lungs and brain. Histopathological analysis revealed cerebral perivascular, lymphohistiocytic infiltrates, focal hemorrhages, meningoencephalitis, and interstitial pneumonia. Disseminated infection of endothelial cells with Orientia in C57BL/6 mice resulted in pathology resembling that of human scrub typhus. The use of this model will allow detailed characterization of the mechanisms of immunity to and pathogenesis of O. tsutsugamushi infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-Propylenoxid, ReagentPlus®, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(±)-Propylenoxid, puriss. p.a., ≥99.5% (GC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
(±)-Propylenoxid, analytical standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.