Merck
  • Identification of two novel brominated contaminants in water samples by ultra-high performance liquid chromatography-Orbitrap Fusion Tribrid mass spectrometer.
Identification of two novel brominated contaminants in water samples by ultra-high performance liquid chromatography-Orbitrap Fusion Tribrid mass spectrometer.
Journal of chromatography. A (2014-12-30)
Aifeng Liu, Guangbo Qu, Chaoli Zhang, Yan Gao, Jianbo Shi, Yuguo Du, Guibin Jiang
ZUSAMMENFASSUNG
Tetrabromobisphenol A mono(2-hydroxyethyl ether) (TBBPA-MHEE) and TBBPA mono(glycidyl ether) (TBBPA-MGE), two impurities of TBBPA derivatives, were hypothesized to be novel brominated contaminants with potential toxicity. However, due to lacking of analytical method and pure standards, their environmental behavior and toxicity have not been studied. Herein we developed a sensitive method based on ultra-high performance liquid chromatography-Orbitrap Fusion Tribrid mass spectrometer (UHPLC-Orbitrap Fusion TMS) for simultaneous detection of TBBPA-MHEE and TBBPA-MGE in water samples. The sample pretreatment method and the experimental conditions of UHPLC and Orbitrap Fusion TMS, were optimized in detail. The instrument detection limits (IDLs) for TBBPA-MHEE and TBBPA-MGE were 0.5pg and 0.6pg, respectively. The method detection limits (MDLs) for TBBPA-MHEE and TBBPA-MGE in river water samples were 0.9 and 0.8ng/L. With the proposed method, we were able to detect TBBPA-MHEE and TBBPA-MGE for the first time in water samples and technical products of TBBPA derivatives. Therefore, UHPLC-Orbitrap Fusion TMS is a simple and effective tool for identification and quantification of novel contaminants in the environment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumhydroxid -Lösung, BioUltra, ~1 M NH3 in H2O (T)
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
3,3′,5,5′-Tetrabrombisphenol A, 97%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
1,3-Propylene sulfite, 99%