Merck
  • Restoration of compact Golgi morphology in advanced prostate cancer enhances susceptibility to galectin-1-induced apoptosis by modifying mucin O-glycan synthesis.
Restoration of compact Golgi morphology in advanced prostate cancer enhances susceptibility to galectin-1-induced apoptosis by modifying mucin O-glycan synthesis.
Molecular cancer research : MCR (2014-08-03)
Armen Petrosyan, Melissa S Holzapfel, David E Muirhead, Pi-Wan Cheng
ZUSAMMENFASSUNG
Prostate cancer progression is associated with upregulation of sialyl-T antigen produced by β-galactoside α-2,3-sialyltransferase-1 (ST3Gal1) but not with core 2-associated polylactosamine despite expression of core 2 N-acetylglucosaminyltransferase-L (C2GnT-L/GCNT1). This property allows androgen-refractory prostate cancer cells to evade galectin-1 (LGALS1)-induced apoptosis, but the mechanism is not known. We have recently reported that Golgi targeting of glycosyltransferases is mediated by golgins: giantin (GOLGB1) for C2GnT-M (GCNT3) and GM130 (GOLGA2)-GRASP65 (GORASP1) or GM130-giantin for core 1 synthase. Here, we show that for Golgi targeting, C2GnT-L also uses giantin exclusively whereas ST3Gal1 uses either giantin or GM130-GRASP65. In addition, the compact Golgi morphology is detected in both androgen-sensitive prostate cancer and normal prostate cells, but fragmented Golgi and mislocalization of C2GnT-L are found in androgen-refractory cells as well as primary prostate tumors (Gleason grade 2-4). Furthermore, failure of giantin monomers to be phosphorylated and dimerized prevents Golgi from forming compact morphology and C2GnT-L from targeting the Golgi. On the other hand, ST3Gal1 reaches the Golgi by an alternate site, GM130-GRASP65. Interestingly, inhibition or knockdown of non-muscle myosin IIA (MYH9) motor protein frees up Rab6a GTPase to promote phosphorylation of giantin by polo-like kinase 3 (PLK3), which is followed by dimerization of giantin assisted by protein disulfide isomerase A3 (PDIA3), and restoration of compact Golgi morphology and targeting of C2GnT-L. Finally, the Golgi relocation of C2GnT-L in androgen-refractory cells results in their increased susceptibility to galectin-1-induced apoptosis by replacing sialyl-T antigen with polylactosamine. This study demonstrates the importance of Golgi morphology and regulation of glycosylation and provides insight into how the Golgi influences cancer progression and metastasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-74, purified immunoglobulin, buffered aqueous solution
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Anti-Myosin IIA, non muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution