Merck
  • Simultaneous screening and identifying four categories of particular flavonoids in the leaves of Murraya exotica L. by HPLC-DAD-ESI-MS-MS.
Simultaneous screening and identifying four categories of particular flavonoids in the leaves of Murraya exotica L. by HPLC-DAD-ESI-MS-MS.
Journal of chromatographic science (2013-01-15)
Jiayu Zhang, Jianqiu Lu, Qian Zhang, Long Dai, Ying Liu, Pengfei Tu, Yanjiang Qiao
ZUSAMMENFASSUNG
Polymethoxylated flavonoids (PMFs), the particular flavonoid subclass in which all or almost all hydroxyls are capped by methylation, have high oral bioavailability and various activities. A sensitive high-performance liquid chromatography-diode array detection-electrospray ionization tandem mass spectrometry (HPLC-DAD-ESI-MS-MS) method was established to screen and identify the PMFs in the leaves of Murraya exotica. Eight PMF standards, including two polymethoxylated flavones, two polymethoxylated flavanones, two polymethoxylated chalcones and two PMF glycosides, were first to be analyzed in positive mode by collision-induced dissociation MS-MS. On the basis of the ESI-MS, characterizations were deduced, and in the results of the extracted ion chromatogram MS-MS experiment, 26 PMFs, including 18 flavones, five flavanones or chalcones and three PMF glycosides, were screened out from the complex extract of the leaves of M. exotica. Among them, 24 PMFs were hydroxylated polymethoxyflavonoids, whereas the rest were all permethoxylated PMFs. This was the first systematic report on the presence of PMFs in the leaves of M. exotica. The results indicated that the established analytical method could be adopted as a rapid, effective technique for the structural characterization of PMFs from the complex extracts of traditional Chinese medicines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Ameisensäure -Lösung, BioUltra, 1.0 M in H2O