Merck
  • Assessment of bacterial resistance to organic solvents using a combined confocal laser scanning and atomic force microscopy (CLSM/AFM).
Assessment of bacterial resistance to organic solvents using a combined confocal laser scanning and atomic force microscopy (CLSM/AFM).
Journal of microbiological methods (2014-09-07)
Maria S Kuyukina, Irena B Ivshina, Irina O Korshunova, Ekaterina V Rubtsova
ZUSAMMENFASSUNG
Using combined confocal laser scanning and atomic force microscopy (CLSM/AFM), bacterial viability under organic solvent stress was assessed at single cell level. Solvent-exposed bacteria stained with the LIVE/DEAD BacLight fluoresced green or red, allowing viable and dead cell discrimination. However, with toluene, butanol and acetonitrile, dually fluorescent cells appeared having compromised cell membranes. Changes in size, surface/volume ratio and roughness were revealed as possible resistance mechanisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Cyclohexan, ACS reagent, ≥99%
Sigma-Aldrich
Cyclohexan, suitable for HPLC, ≥99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
1-Butanol, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Decan, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Cyclohexan, anhydrous, 99.5%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
1-Butanol, Molecular Biology, ≥99%
Supelco
Toluol, analytical standard
Sigma-Aldrich
Decan, ≥95%
Sigma-Aldrich
Decan, anhydrous, ≥99%
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cyclohexan, ACS reagent, ≥99%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Decan, analytical standard