Merck
  • Quantification of filamentation by uropathogenic Escherichia coli during experimental bladder cell infection by using semi-automated image analysis.
Quantification of filamentation by uropathogenic Escherichia coli during experimental bladder cell infection by using semi-automated image analysis.
Journal of microbiological methods (2014-12-30)
Kasper Klein, Yaseelan Palarasah, Hans Jørn Kolmos, Jakob Møller-Jensen, Thomas Emil Andersen
ZUSAMMENFASSUNG
Several rod-shaped pathogens including Escherichia coli, Salmonella spp. and Klebsiella pneumonia are capable of adopting highly filamentous cell shapes under certain circumstances. This phenomenon occurs as a result of continued cell elongation during growth without the usual septation into single rod-shaped cells. Evidence has emerged over the past decade suggesting that this morphological transformation is controlled and reversible and provides selective advantages under certain growth conditions, such as during infection in humans. In order to identify the factors which induce filamentation of bacterial pathogens and study the advantages of bacterial morphological plasticity, methods are needed to accurately quantify changes in bacterial cell shape. In this study, we present a method for quantification of bacterial filamentation based on automatic detection and measurement of bacterial units in focus-stacked microscopy images. Used in combination with a flow-chamber based in vitro cystitis model, we study the factors involved in filament formation by uropathogenic E. coli (UPEC) during infection. The influence of substratum surface, intracellular proliferation and flow media on UPEC filamentation is evaluated. We show that reversible UPEC filamentation during cystitis is not dependent on intracellular infection, which previous studies have suggested. Instead, we find that filamentation can be induced by contact with surfaces, both biological and artificial. Lastly our data indicate that UPEC filamentation is induced by trace-amounts of specific components in urine, rather than being a generic stress-response to high urine salt concentrations. The study shows that the combined methodology is generally useful for investigation of bacterial morphological transitions during cell infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammoniumchlorid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Ammoniumchlorid, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Harnsäure, ≥99%, crystalline
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Milchsäure, meets USP testing specifications
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®