Merck
Medical-grade collagen peptide in injectables provides antioxidant protection.
Pharmaceutical development and technology (2013-11-30)
Kyo Kobayashi, Yojiro Maehata, Yasue Okada, Masashi Kusubata, Shunji Hattori, Keisuke Tanaka, Chihiro Miyamoto, Fumihiko Yoshino, Ayaka Yoshida, Fumiaki Tokutomi, Satoko Wada-Takahashi, Tomoko Komatsu, Takero Otsuka, Shun-Suke Takahashi, Masaichi-Chang-Il Lee
ZUSAMMENFASSUNG
Medical-grade collagen peptide is used as an additive agent in pharmaceutical formulations; however, it is unknown as to whether the compound exerts antioxidant effects in vitro. In this study, we evaluated the antioxidant effects of medical-grade collagen peptide on reactive oxygen species such as hydroxyl radical, superoxide anion radical and singlet oxygen using electron spin resonance and spin trapping. We confirmed that medical-grade collagen peptide directly inhibited hydroxyl radical generated by the Fenton reaction or by ultraviolet irradiation of hydrogen peroxide, and singlet oxygen. In addition, an antioxidant effect of medical-grade collagen peptide on singlet oxygen was observed in peptide fractions 12-22. The total amount of antioxidant amino acids (Gly, Hyp, Glu, Ala, Cys, Met and His) constituted more than half of the total amino acids in these fractions. These results suggest that the observed antioxidant properties of medical-grade collagen peptide are due to the compound containing antioxidant amino acids. Medical-grade collagen peptide, which is used in pharmaceuticals, and especially in injectables, could provide useful antioxidant properties to protect the active ingredient from oxidation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Wasserstoffperoxid -Lösung, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, ReagentPlus®, ≥99%
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, ACS reagent, ≥99.0%
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 50 wt. % in H2O, stabilized
Sigma-Aldrich
L-Histidin, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumsalicylat, ReagentPlus®, ≥99.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Histidin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidin, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Millipore
Wasserstoffperoxid -Lösung, 3%, suitable for microbiology
Supelco
Wasserstoffperoxid -Lösung, ≥30%, for trace analysis
Sigma-Aldrich
Eisen(II)-sulfat Heptahydrat, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
SAFC
L-Histidin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications