Merck
  • Autoreactive T cells induce necrosis and not BCL-2-regulated or death receptor-mediated apoptosis or RIPK3-dependent necroptosis of transplanted islets in a mouse model of type 1 diabetes.
Autoreactive T cells induce necrosis and not BCL-2-regulated or death receptor-mediated apoptosis or RIPK3-dependent necroptosis of transplanted islets in a mouse model of type 1 diabetes.
Diabetologia (2014-10-11)
Yuxing Zhao, Nicholas A Scott, Stacey Fynch, Lorraine Elkerbout, W Wei-Lynn Wong, Kylie D Mason, Andreas Strasser, David C Huang, Thomas W H Kay, Helen E Thomas
ZUSAMMENFASSUNG
Type 1 diabetes results from T cell-mediated destruction of pancreatic beta cells. The mechanisms of beta cell destruction in vivo, however, remain unclear. We aimed to test the relative roles of the main cell death pathways: apoptosis, necrosis and necroptosis, in beta cell death in the development of CD4(+) T cell-mediated autoimmune diabetes. We altered expression levels of critical cell death proteins in mouse islets and tested their ability to survive CD4(+) T cell-mediated attack using an in vivo graft model. Loss of the B cell leukaemia/lymphoma 2 (BCL-2) homology domain 3-only proteins BIM, PUMA or BID did not protect beta cells from this death. Overexpression of the anti-apoptotic protein BCL-2 or combined deficiency of the pro-apoptotic multi-BCL2 homology domain proteins BAX and BAK also failed to prevent beta cell destruction. Furthermore, loss of function of the death receptor Fas or its essential downstream signalling molecule Fas-associated death domain (FADD) in islets was also not protective. Using electron microscopy we observed that dying beta cells showed features of necrosis. However, islets deficient in receptor-interacting serine/threonine protein kinase 3 (RIPK3), a critical initiator of necroptosis, were still normally susceptible to CD4(+) T cell-mediated destruction. Remarkably, simultaneous inhibition of apoptosis and necroptosis by combining loss of RIPK3 and overexpression of BCL-2 in islets did not protect them against immune attack either. Collectively, our data indicate that beta cells die by necrosis in autoimmune diabetes and that the programmed cell death pathways apoptosis and necroptosis are both dispensable for this process.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
D-Luciferin, synthetic
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)
Isofluran, European Pharmacopoeia (EP) Reference Standard