Merck
Oxidative stress status in patients with acute urticaria.
Cutaneous and ocular toxicology (2013-07-12)
Göknur Kalkan, Havva Yıldız Seçkin, Fazilet Duygu, Ali Akbaş, Hüseyin Ozyurt, Mehmet Sahin
ZUSAMMENFASSUNG
The conflicting information related to oxidative stress status in patients with chronic idiopathic urticaria has been reported in several studies. However, the association between acute urticaria (AU) and oxidative stress has not been investigated exhaustively. To evaluate the role of the oxidative stress in the patients with AU by determining the oxidant/antioxidant activity in AU and to establish its clinical significance. About 50 patients with AU, (10 males, 40 females) and 30 unrelated healthy controls (4 males, 26 females) were enrolled into the study. The activity of the antioxidant enzymes copper-zinc superoxide dismutase (Cu-Zn-SOD), glutathione peroxidase (GSH-Px) and catalase, and the levels of malondialdehyde (MDA), serum NO and protein carbonyls levels in the plasma were measured spectrophotometrically at samples. A statistically significant increase was observed in serum Cu-ZnSOD activities of the patients when compared with that of the controls (p < 0.001), while a statistically significant decrease was observed in GSH-Px activities of the patients according to the controls (p = 0.002). Serum MDA and NO levels were significantly higher in patients with AU when compared with control group (p < 0.001 for both of them). The levels of protein carbonyls were significantly lower in patients with AU when compared with control group (p < 0.001). It seems there is an oxidative burden in the patients with AU. Cutaneous oxidative stress may play a role in pathogenesis of the disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 50% in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natronlauge, 5.0 M
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natronlauge, 49-51% in water, eluent for IC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O
Preise und Verfügbarkeit sind derzeit nicht verfügbar.