Merck
Influence of kinin peptides on monocyte-endothelial cell adhesion.
Journal of cellular biochemistry (2014-06-14)
Ibeth Guevara-Lora, Krystyna Stalinska, Bartlomiej Augustynek, Anna Labedz-Maslowska
ZUSAMMENFASSUNG
Adhesion of leukocytes to vascular endothelium in response to proinflammatory mediators is an important component of the overall inflammatory reaction. In the current work, we used a retinoic acid-differentiated human promonocytic cell line, U937 and a human microvascular endothelial cell line, HMEC-1 to analyze the effect of the potent pro-inflammatory bradykinin-related peptides (kinins) on cell adhesion. Bradykinin (BK) and kinin metabolites without the C-terminal arginine residue enhanced adhesion of the monocyte-like cells to fibronectin and to the HMEC-1 cells. Expression of adhesion proteins on the surface of both cell types was altered by the kinin peptides. In the monocyte-like cells, expression of CD11b, a subunit of Mac-1 integrin, was significantly increased whilst in the endothelial cells, a strong increase in the production of intercellular adhesion molecule 1 (ICAM-1) was observed. The positive bradykinin-induced effect on the cell-cell interaction was reversed by a carboxypeptidase inhibitor (MGTA), hence we suspected a significant role of the des-Arg kinin metabolites, which acted through the kinin receptor type 1. Indeed, the expression of this receptor was up-regulated not only by agonists but also by interferon-γ and bradykinin. Kinin peptides also regulated signal transducer and activator of transcription proteins (STATs) activated by cytokines. Taken together, the above observations support our hypothesis that kinins stimulate monocyte adhesion to the vessel wall, especially during pathological states of the circulatory system accompanied by proinflammatory cytokine release.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
HOE 140, ≥94%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Supelco
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
USP
Captopril, United States Pharmacopeia (USP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Hydrocortison, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Captopril, meets USP testing specifications
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Hydrocortison, British Pharmacopoeia (BP) Assay Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Captopril, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Captopril für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Preise und Verfügbarkeit sind derzeit nicht verfügbar.