Merck
PCR-free blood group genotyping using a nanobiosensor.
Vox sanguinis (2014-12-04)
D Brouard, O Ratelle, J Perreault, D Boudreau, M St-Louis
ZUSAMMENFASSUNG
The last two decades have seen major developments in genotyping assays to facilitate the procurement of red blood cell units to alloimmunized patients. To make genotyping faster, simpler and less costly, a nanotechnology approach based on metal/silica fluorescent nanoparticles and a polymer-based hybridization optical transducer was designed. The objectives of this study were (1) to verify whether this nanobiosensor has the ability to discriminate single nucleotide polymorphisms in non-amplified genomic DNA and (2) to establish whether the signal generated by the nanobiosensor is sufficiently intense to be detected by standard flow cytometry. Silver-core silica-shell fluorescent nanoparticles (Ag@SiO₂) were prepared, and amine-modified DNA probes were grafted to their surface. A cationic conjugated polymer was electrostatically bound to the surface probes to become optically active upon hybridization with a target. Two nanobiosensor formulations specific to DO*01 and DO*02 alleles were prepared. DNA was extracted from whole blood and mixed with the nanobiosensor for hybridization. The nanobiosensor fluorescence was measured by flow cytometry. Nine volunteers were typed for Dombrock blood group antigens DO*01 and DO*02. A statistically significant increase in the optical transduction signal was observed for sequence-specific samples. All nine genotypes were correctly identified when compared to standardized PCR assays. The nanobiosensor provides rapid and simple genotyping of blood group antigens from unamplified genomic DNA and can be measured using standard flow cytometers. This PCR-free approach could be applied to any known genetic polymorphism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Bromphenolblau, Sulton Form, ACS reagent
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Bromphenolblau, titration: suitable
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder