Merck
  • Potential inhibitory effect of herbal medicines on rat hepatic cytochrome P450 2D gene expression and metabolic activity.
Potential inhibitory effect of herbal medicines on rat hepatic cytochrome P450 2D gene expression and metabolic activity.
Die Pharmazie (2015-05-20)
F I Al-Jenoobi, H M Korashy, A Ahad, M Raish, A M Al-Mohizea, M A Alam, S A Al-Suwayeh, K M Alkharfy
ZUSAMMENFASSUNG
The aim of current study was to investigate the effect of some commonly used medicinal herbs on the regulation of rat CYP2D gene expression and its metabolic activity. Wistar albino rats were treated for seven consecutive days with selected doses of five commonly used herbs (Trigonella foenum-graecum, Ferula asafoetida, Nigella sativa, Commiphora myrrha and Lepidium sativum). Thereafter, rat livers were harvested and CYP2D mRNA levels were determined by RT-PCR. The metabolic activity of CYP2D was performed on rat hepatic microsomes using dextromethorphan as specific substrate. All investigated herbs produced inhibition of CYP2D mRNA expression and metabolic activity. The inhibitory potential of investigated herbs on rat CYP2D mRNA was in the following order: Commiphora myrrha > Nigella sativa > Lepidium sativum > Trigonella foenum-graecum > Ferula asafoetida. Whereas, the inhibitory potential of investigated herbs on CYP2D mediated enzyme metabolic activity was found in following order: Nigella sativa > Lepidium sativum > Trigonella foenum-graecum > Commiphora myrrha > Ferula asafoetida. The current study shows that only used herbs reduce CYP2D activity in rat liver microsomes at the transcriptional levels. Such effects could lead to undesirable pharmacological effects of clinically used low therapeutic index CYP2D substrate drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Supelco
Kaliumphosphat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumphosphat, tested according to Ph. Eur., anhydrous
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
USP
Monobasisches Kaliumphosphat, United States Pharmacopeia (USP) Reference Standard
USP
Lösungsmittelrückstände Klasse 2 - Acetonitril, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitril, Pharmaceutical Secondary Standard; Certified Reference Material